Gene Transcripts

Gene Symbol
Fam207a
Entrez Gene ID
108707
Gene Name
family with sequence similarity 207, member A
Gene Aliases
1810008A18Rik, AI303072
Chromosome Location
Chr. 10: 77486655 - 77537969 on Build GRCm38
UniGene ID
Mm.259547
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133998.3 NP_598759.1 5, 6 617
GenBank mRNA AF391115.1 - 5, 6 590
AK076516.1 - 5, 6 585
AK088306.1 - 5, 6 585
AK160157.1 - 5, 6 488
BC034216.1 - 5, 6 563 5008732

View Full Product Details