Gene Transcripts
- Gene Symbol
- Olfr23
- Entrez Gene ID
- 18321
- Gene Name
- olfactory receptor 23
- Gene Aliases
- MOR135-27, MTPCR50
- Chromosome Location
- Chr. 11: 73930360 - 73941225 on Build GRCm38
- UniGene ID
- Mm.246529
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010970.1 |
NP_035100.1
|
1 |
767 |
|
GenBank mRNA |
X89688.1 |
- |
1 |
393 |
|
View Full Product Details