Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153098.3 | NP_694738.1 | 4 | 636 | |
GenBank mRNA | AK029247.1 | - | 4 | 411 | |
AK036799.1 | - | 4 | 396 | ||
AY083458.1 | - | 4 | 642 | ||
BC052443.1 | - | 4 | 404 | 6406841 | |
DQ832284.1 | - | 4 | 642 |