Gene Transcripts

Gene Symbol
Arl9
Entrez Gene ID
384185
Gene Name
ADP-ribosylation factor-like 9
Gene Aliases
1700110A09Rik
Chromosome Location
Chr. 5: 77002805 - 77010959 on Build GRCm38
UniGene ID
Mm.451858
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206935.1 NP_996818.1 2 99

Gene Symbol
1700112J05Rik
Entrez Gene ID
68246
Gene Name
RIKEN cDNA 1700112J05 gene
Gene Aliases
1700110A09Rik
Chromosome Location
Chr. 5: 77006442 - 77018863 on Build GRCm38
UniGene ID
Mm.319633
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK018948.1 - 4 714

View Full Product Details