Gene Transcripts

Gene Symbol
Rnf168
Entrez Gene ID
70238
Gene Name
ring finger protein 168
Gene Aliases
3110001H15Rik
Chromosome Location
Chr. 16: 32277461 - 32301442 on Build GRCm38
UniGene ID
Mm.228263
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027355.2 NP_081631.2 6 1773
GenBank mRNA BC046815.1 - 6 1745

View Full Product Details