Gene Transcripts

Gene Symbol
Slc22a15
Entrez Gene ID
310732
Gene Name
solute carrier family 22, member 15
Gene Aliases
Chromosome Location
Chr. 2: 204281638 - 204344026 on Build Rnor_6.0
UniGene ID
Rn.46389
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107707.1 NP_001101177.1 6 981
XM_006233050.2 XP_006233112.1 5 3375

View Full Product Details