Gene Transcripts

Gene Symbol
Ddx17
Entrez Gene ID
315133
Gene Name
DEAD (Asp-Glu-Ala-Asp) box helicase 17
Gene Aliases
Chromosome Location
Chr. 7: 120761843 - 120765012 on Build Rnor_6.0
UniGene ID
Rn.224752
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015018.1 NP_001015018.1 2 530
GenBank mRNA BC092629.1 - 2 530

View Full Product Details