Gene Transcripts

Gene Symbol
Ebf2
Entrez Gene ID
361060
Gene Name
early B-cell factor 2
Gene Aliases
Chromosome Location
Chr. 15: 43905099 - 44103742 on Build Rnor_6.0
UniGene ID
Rn.137334
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108383.1 NP_001101853.1 16 2645
XM_006252133.2 XP_006252195.1 15 2222

View Full Product Details