Gene Transcripts

Gene Symbol
Fgf16
Entrez Gene ID
60464
Gene Name
fibroblast growth factor 16
Gene Aliases
Chromosome Location
Chr. X: 76786728 - 76796311 on Build Rnor_6.0
UniGene ID
Rn.44348
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021867.3 NP_068639.1 2 332
GenBank mRNA AB002561.1 - 2 332
BC085738.1 - 606 7189721

View Full Product Details