Gene Transcripts

Gene Symbol
Bbox1
Entrez Gene ID
64564
Gene Name
butyrobetaine (gamma), 2-oxoglutarate dioxygenase (gamma-butyrobetaine hydroxylase) 1
Gene Aliases
Bbh, Bbox
Chromosome Location
Chr. 3: 101427052 - 101465995 on Build Rnor_6.0
UniGene ID
Rn.16333
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022629.1 NP_072151.1 7 1074
GenBank mRNA AF160958.1 - 7 1074
FQ210746.1 - 1166

View Full Product Details