Gene Transcripts

Gene Symbol
Mkln1
Entrez Gene ID
83536
Gene Name
muskelin 1, intracellular mediator containing kelch motifs
Gene Aliases
Chromosome Location
Chr. 4: 58693384 - 58817924 on Build Rnor_6.0
UniGene ID
Rn.6383
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031359.1 NP_112649.1 10 1051
GenBank mRNA AB046442.1 - 10 1051

View Full Product Details