Gene Transcripts

Gene Symbol
Prpf38a
Entrez Gene ID
298374
Gene Name
pre-mRNA processing factor 38A
Gene Aliases
RGD1311129
Chromosome Location
Chr. 5: 128174835 - 128186532 on Build Rnor_6.0
UniGene ID
Rn.18104
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106672.1 NP_001100142.1 8, 9 1059
GenBank mRNA FQ221356.1 - 6, 7 465

View Full Product Details