Gene Transcripts

Gene Symbol
Antxr1
Entrez Gene ID
362393
Gene Name
anthrax toxin receptor 1
Gene Aliases
Chromosome Location
Chr. 4: 118946267 - 119131202 on Build Rnor_6.0
UniGene ID
Rn.41192
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044249.2 NP_001037714.1 7, 8 946
GenBank mRNA AY754025.1 - 6, 7 377
BC131853.1 - 7, 8 946 7320366
DQ789143.1 - 7, 8 595

View Full Product Details