Gene Transcripts

Gene Symbol
Rpl4
Entrez Gene ID
64302
Gene Name
ribosomal protein L4
Gene Aliases
Chromosome Location
Chr. 8: 69121682 - 69126805 on Build Rnor_6.0
UniGene ID
Rn.1133
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022510.1 NP_071955.1 2 205
GenBank mRNA BC081801.1 - 2 176
FQ209448.1 - 1 59
FQ209532.1 - 1 59
FQ212557.1 - 1 21
FQ212566.1 - 1 59
FQ212656.1 - 1 59
FQ212896.1 - 1 59
FQ213045.1 - 1 59
FQ213454.1 - 2 210
FQ215017.1 - 1 15
FQ215042.1 - 1 59
FQ215264.1 - 1 59
FQ215283.1 - 1 59
FQ215533.1 - 1 59
FQ215586.1 - 1 59
FQ215649.1 - 1 59
FQ216342.1 - 1 59
FQ216573.1 - 1 59
FQ216754.1 - 1 59
FQ216792.1 - 1 59
FQ218195.1 - 1 59
FQ219327.1 - 1 59
FQ219561.1 - 1 59
FQ219701.1 - 1 59
FQ219716.1 - 1 59
FQ220231.1 - 1 59
FQ221130.1 - 1 21
FQ221674.1 - 1 59
FQ226216.1 - 1 59
FQ228709.1 - 1 59
FQ228734.1 - 2 210
FQ228797.1 - 1 59
FQ228847.1 - 1 59
FQ228910.1 - 1 59
FQ228929.1 - 1 59
FQ228930.1 - 1 59
FQ229086.1 - 1 59
FQ229087.1 - 1 59
FQ229116.1 - 1 59
FQ229849.1 - 1 18
FQ230368.1 - 1 59
FQ231653.1 - 1 59
FQ232105.1 - 1 59
FQ232562.1 - 1 59
FQ232771.1 - 1 59
FQ233898.1 - 1 59
FQ234293.1 - 1 59
FQ234521.1 - 1 59
FQ234619.1 - 1 59
X82180.1 - 2 205

View Full Product Details