Gene Transcripts

Gene Symbol
Dpysl5
Entrez Gene ID
65208
Gene Name
dihydropyrimidinase-like 5
Gene Aliases
Crmp5, Ulip6
Chromosome Location
Chr. 6: 26939697 - 27024129 on Build Rnor_6.0
UniGene ID
Rn.44871
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023023.1 NP_075412.1 2 232
GenBank mRNA AB029432.1 - 1 109
AJ131436.1 - 2 232

View Full Product Details