Gene Transcripts

Gene Symbol
PPP2CB
Entrez Gene ID
5516
Gene Name
protein phosphatase 2 catalytic subunit beta
Gene Aliases
PP2Abeta, PP2CB
Chromosome Location
Chr.8: 30785610 - 30812836 on Build GRCh38
Uni Gene
Hs.491440
Interrogated Sequence Translated Protein
RefSeq NM_001009552.1 NP_001009552.1
GenBank mRNA AF444006.1 -
AK091875.1 -
BC012022.1 -
BG716837.1 -
BT019526.1 -
CA419001.1 -
CF594136.1 -
CR541747.1 -
ENST00000221138.0 -
J03805.1 -
X12656.1 -

View Full Product Details