Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001009552.1 | NP_001009552.1 |
GenBank mRNA | AF444006.1 | - |
AK091875.1 | - | |
BC012022.1 | - | |
BG716837.1 | - | |
BT019526.1 | - | |
CA419001.1 | - | |
CF594136.1 | - | |
CR541747.1 | - | |
ENST00000221138.0 | - | |
J03805.1 | - | |
X12656.1 | - |