Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001316332.1 | NP_001303261.1 |
NM_002756.4 | NP_002747.2 | |
NM_145109.2 | NP_659731.1 | |
XM_005256723.2 | XP_005256780.1 | |
XM_011523958.2 | XP_011522260.1 | |
XM_011523959.1 | XP_011522261.1 | |
XM_017024857.1 | XP_016880346.1 | |
XM_017024858.1 | XP_016880347.1 | |
XM_017024859.1 | XP_016880348.1 | |
GenBank mRNA | AB451226.1 | - |
AB451349.1 | - | |
AI758302.1 | - | |
AK093838.1 | - | |
AK223490.1 | - | |
AK315505.1 | - | |
BC032478.1 | - | |
BM836867.1 | - | |
CR536514.1 | - | |
CR541793.1 | - | |
D87116.1 | - | |
DA131175.1 | - | |
DA987047.1 | - | |
ENST00000361818.0 | - | |
GQ225578.1 | - | |
L36719.1 | - | |
U66839.1 | - |