Gene Transcripts

Gene Symbol
Map2k3
Entrez Gene ID
26397
Gene Name
mitogen-activated protein kinase kinase 3
Gene Aliases
AW212142, MEK3, MKK3, Prkmk3, mMKK3b
Chromosome Location
Chr.11: 60932057 - 60952803 on Build GRCm38
Uni Gene
Mm.18494
Interrogated Sequence Translated Protein
RefSeq NM_008928.4 NP_032954.1
GenBank mRNA AK008141.1 -
AK011002.1 -
AK053378.1 -
AK150067.1 -
AK159379.1 -
AK166255.1 -
AK166507.1 -
AK167209.1 -
AK167442.1 -
AK168710.1 -
AK176968.1 -
AK179609.1 -
AK181928.1 -
AK185191.1 -
AK197832.1 -
AK219340.1 -
BC007467.1 -
D87115.1 -
X93150.1 -

View Full Product Details