Gene Transcripts

Gene Symbol
Ppp2cb
Entrez Gene ID
19053
Gene Name
protein phosphatase 2 (formerly 2A), catalytic subunit, beta isoform
Gene Aliases
AI115466, D8Ertd766e, PP2Ac
Chromosome Location
Chr.8: 33599621 - 33619804 on Build GRCm38
Uni Gene
Mm.288765
Interrogated Sequence Translated Protein
RefSeq NM_017374.3 NP_059070.1
GenBank mRNA AA220297.1 -
AK090311.1 -
BC019161.1 -
BC049594.1 -
BC058582.1 -
BE847789.1 -
CK619614.1 -
Z67746.1 -

View Full Product Details