Gene Transcripts

Gene Symbol
Alg12
Entrez Gene ID
223774
Gene Name
asparagine-linked glycosylation 12 (alpha-1,6-mannosyltransferase)
Gene Aliases
ECM39
Chromosome Location
Chr.15: 88805243 - 88819318 on Build GRCm38
Uni Gene
Mm.41512
Interrogated Sequence Translated Protein
RefSeq NM_001142357.1 NP_001135829.1
NM_145477.2 NP_663452.1
GenBank mRNA AJ429133.1 -
AK053357.1 -
AK142322.1 -
AK179946.1 -
AK187237.1 -
AK209258.1 -
BB480728.1 -
BC021379.1 -

View Full Product Details