Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_008928.4 | NP_032954.1 |
GenBank mRNA | AK008141.1 | - |
AK011002.1 | - | |
AK053378.1 | - | |
AK150067.1 | - | |
AK159379.1 | - | |
AK166255.1 | - | |
AK166507.1 | - | |
AK167209.1 | - | |
AK167442.1 | - | |
AK168710.1 | - | |
AK176968.1 | - | |
AK179609.1 | - | |
AK181928.1 | - | |
AK185191.1 | - | |
AK197832.1 | - | |
AK219340.1 | - | |
BC007467.1 | - | |
D87115.1 | - | |
X93150.1 | - |