Gene Transcripts

Gene Symbol
Ntf4
Entrez Gene ID
25730
Gene Name
neurotrophin 4
Gene Aliases
NT4P, Ntf5
Chromosome Location
Chr.1: 101405314 - 101408145 on Build Rnor_6.0
Uni Gene
Rn.44225
Interrogated Sequence Translated Protein
RefSeq NM_013184.3 NP_037316.2
GenBank mRNA S69323.1 -

View Full Product Details