Gene Transcripts

Gene Symbol
ADAM6A
Entrez Gene ID
100303770
Gene Name
cellular disintegrin ADAM 6e
Gene Aliases
tMDCIVe
Chromosome Location
Chr.17: 82311883 - 82314286 on Build OryCun2.0
Uni Gene
Ocu.2200
Interrogated Sequence Translated Protein
RefSeq NM_001164866.1 NP_001158338.1
GenBank mRNA U82751.1 -

View Full Product Details