Gene Transcripts

Gene Symbol
1700028J19Rik
Entrez Gene ID
70004
Gene Name
RIKEN cDNA 1700028J19 gene
Gene Aliases
Chromosome Location
Chr. 7: 44229929 - 44236122 on Build GRCm38
UniGene ID
Mm.158317
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_029436.1 3, 4 399
GenBank mRNA AK006462.1 - 3, 4 400
BC049653.1 - 3, 4 365
BC100477.1 - 3, 4 381

View Full Product Details