Gene Transcripts

Gene Symbol
4933407K13Rik
Entrez Gene ID
74396
Gene Name
RIKEN cDNA 4933407K13 gene
Gene Aliases
A130080L05Rik
Chromosome Location
Chr. X: 75725458 - 75764699 on Build GRCm38
UniGene ID
Mm.162493
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_029443.1 9 1836
GenBank mRNA AK016728.1 - 5 918
AK030939.1 - 10 1837
AK036260.1 - 1 1757
AK053637.1 - 9 1950

View Full Product Details