Gene Transcripts
- Gene Symbol
- Btnl5-ps
- Entrez Gene ID
- 81497
- Gene Name
- butyrophilin-like 5, pseudogene
- Gene Aliases
- Btnl5, ENSMUSG00000073420, Gm15330, NG12, OTTMUSG00000020237
- Chromosome Location
- Chr. 17: 34487405 - 34497429 on Build GRCm38
- UniGene ID
- Mm.484231
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_004051.1 |
|
9 |
2377 |
|
View Full Product Details