Gene Transcripts
- Gene Symbol
- ST7-AS2
- Entrez Gene ID
- 93654
- Gene Name
- ST7 antisense RNA 2
- Gene Aliases
- ST7AS2, ST7OT2
- Chromosome Location
- Chr. 7: 117072072 - 117145592 on Build GRCh38
- UniGene ID
- Hs.628891
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002331.3 |
|
|
2, 3 |
289 |
|
GenBank mRNA |
AF400045.1 |
- |
LIT1899 ST7OT2 |
2, 3 |
279 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX097853.1 |
- |
2, 3 |
256 |
|
Chr. 7 - 117112518 - 117145559 [-] |
View Full Product Details