Gene Transcripts

Gene Symbol
MOBP
Entrez Gene ID
4336
Gene Name
myelin-associated oligodendrocyte basic protein
Gene Aliases
Chromosome Location
Chr. 3: 39467573 - 39529497 on Build GRCh38
UniGene ID
Hs.121333
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003090.2 6 2318
NR_103504.1 7 2660
NR_103505.1 7 2698
GenBank mRNA BC022471.1 - 6 2213

View Full Product Details