Gene Transcripts

Gene Symbol
LINC01142
Entrez Gene ID
284688
Gene Name
long intergenic non-protein coding RNA 1142
Gene Aliases
Chromosome Location
Chr. 1: 170271405 - 170284208 on Build GRCh38
UniGene ID
Hs.135668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026957.1 3 509
GenBank mRNA BC033381.2 - 2 439
DB058759.1 - 3 459

View Full Product Details