Gene Transcripts
- Gene Symbol
- LINC00152
- Entrez Gene ID
- 112597
- Gene Name
- long intergenic non-protein coding RNA 152
- Gene Aliases
- C2orf59, NCRNA00152
- Chromosome Location
- Chr. 2: 87455455 - 87521518 on Build GRCh38
- UniGene ID
- Hs.652166
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC070202.1 |
- |
|
27 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CD639006.1 |
- |
1 |
30 |
|
Chr. 2 - 87509214 - 87521518 [+] |
View Full Product Details