Gene Transcripts
- Gene Symbol
- LOC102724659
- Entrez Gene ID
- 102724659
- Gene Name
- uncharacterized LOC102724659
- Gene Aliases
- Chromosome Location
- Chr. 1: 11777077 - 11779619 on Build GRCh38
- UniGene ID
- Hs.737635
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_126000.1 |
|
3 |
1817 |
|
GenBank mRNA |
BC034298.1 |
- |
3 |
1816 |
|
View Full Product Details