Gene Transcripts

Gene Symbol
GLB1L3
Entrez Gene ID
112937
Gene Name
galactosidase beta 1 like 3
Gene Aliases
Chromosome Location
Chr. 11: 134275447 - 134321364 on Build GRCh38
UniGene ID
Hs.721382
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011542567.2 XP_011540869.1 19 2415


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CR608608.1 - 2 90 Chr. 11 - 134318747 - 134321301 [+]

View Full Product Details