Gene Transcripts
- Gene Symbol
- GLB1L3
- Entrez Gene ID
- 112937
- Gene Name
- galactosidase beta 1 like 3
- Gene Aliases
- Chromosome Location
- Chr. 11: 134275447 - 134321364 on Build GRCh38
- UniGene ID
- Hs.721382
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX439455.2 |
- |
2 |
830 |
|
Chr. 11 - 134318751 - 134321285 [+] |
GenBankmRNA |
CR608608.1 |
- |
2 |
834 |
|
Chr. 11 - 134318747 - 134321301 [+] |
View Full Product Details