Gene Transcripts
- Gene Symbol
- Gm29154
- Entrez Gene ID
- 102635534
- Gene Name
- predicted gene 29154
- Gene Aliases
- Chromosome Location
- Chr. 9: 103919392 - 103960323 on Build GRCm38
- UniGene ID
- Mm.65435
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK131742.1 |
- |
4 |
661 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY706784.1 |
- |
4 |
661 |
|
Chr. 9 - 103919501 - 103959421 [-] |
View Full Product Details