Gene Transcripts
- Gene Symbol
- P2rx3
- Entrez Gene ID
- 228139
- Gene Name
- purinergic receptor P2X, ligand-gated ion channel, 3
- Gene Aliases
- 4930513E20Rik, P2X3
- Chromosome Location
- Chr. 2: 84996396 - 85037029 on Build GRCm38
- UniGene ID
- Mm.440375
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK019679.1 |
- |
2 |
2094 |
|
View Full Product Details