Gene Transcripts
- Gene Symbol
- Gm32754
- Entrez Gene ID
- 102635403
- Gene Name
- predicted gene, 32754
- Gene Aliases
- Chromosome Location
- Chr. 3: 140909907 - 140917545 on Build GRCm38
- UniGene ID
- Mm.108003
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK076796.1 |
- |
|
470 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB013538.2 |
- |
4 |
290 |
|
Chr. 3 - 140904998 - 140917352 [-] |
View Full Product Details