Gene Transcripts
- Gene Symbol
- Gm32736
- Entrez Gene ID
- 102635379
- Gene Name
- predicted gene, 32736
- Gene Aliases
- Chromosome Location
- Chr. 5: 106094943 - 106112121 on Build GRCm38
- UniGene ID
- Mm.137042
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK053481.1 |
- |
2 |
195 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB665067.1 |
- |
2 |
195 |
|
Chr. 5 - 106095091 - 106101486 [+] |
View Full Product Details