Gene Transcripts
- Gene Symbol
- JARID2-AS1
- Entrez Gene ID
- 100506681
- Gene Name
- JARID2 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 15247815 - 15248696 on Build GRCh38
- UniGene ID
- Hs.664092
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_120502.1 |
|
2 |
482 |
|
GenBank mRNA |
BC033996.1 |
- |
2 |
443 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000441978.1 |
- |
2 |
420 |
|
Chr. 6 - 15247815 - 15248634 [-] |
View Full Product Details