Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289798.1 | NP_001276727.1 | 13 | 1253 | |
NM_001289799.1 | NP_001276728.1 | 13 | 1249 | ||
NM_145558.2 | NP_663533.1 | 13 | 1206 | ||
GenBank mRNA | AK033462.1 | - | 13 | 1188 | |
AK076814.1 | - | 13 | 1169 | ||
AK083164.1 | - | 13 | 1214 | ||
AK083767.1 | - | 13 | 1169 | ||
AK150889.1 | - | 13 | 1168 | ||
AK168594.1 | - | 12 | 1033 | ||
AK169637.1 | - | 13 | 1182 | ||
BC005585.1 | - | 12 | 1137 | 3158015 |