Gene Transcripts

Gene Symbol
Mtnr1b
Entrez Gene ID
244701
Gene Name
melatonin receptor 1B
Gene Aliases
Mel-1B-R, Mel1b, Mt2
Chromosome Location
Chr. 9: 15862614 - 15874556 on Build GRCm38
UniGene ID
Mm.222631
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145712.2 NP_663758.2 1, 2 320
GenBank mRNA AB377276.1 - 320
AY145850.1 - 1, 2 224
BC104324.1 - 1, 2 224 40044921
BC104325.1 - 1, 2 224 40044928

View Full Product Details