Gene Transcripts

Gene Symbol
Dcpp3
Entrez Gene ID
620253
Gene Name
demilune cell and parotid protein 3
Gene Aliases
Dcpp-3, EG620253, SPT-3
Chromosome Location
Chr. 17: 23916698 - 23919441 on Build GRCm38
UniGene ID
Mm.390964
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077633.1 NP_001071101.1 4 489

Gene Symbol
Dcpp1
Entrez Gene ID
13184
Gene Name
demilune cell and parotid protein 1
Gene Aliases
BC005655, C86550, Dcpp, Dcpp-1, p20
Chromosome Location
Chr. 17: 23880876 - 23882853 on Build GRCm38
UniGene ID
Mm.371556
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019910.2 NP_064294.2 4 496
GenBank mRNA BC005655.1 - 493
BC024627.1 - 470 4925312
BC045522.1 - 4 480 4190591
BC090399.1 - 470 4168483
DQ237936.1 - 488
S76879.1 - 529
U03711.1 - 498

Gene Symbol
Dcpp2
Entrez Gene ID
630537
Gene Name
demilune cell and parotid protein 2
Gene Aliases
Dcpp-2, EG630537, SPT-2
Chromosome Location
Chr. 17: 23898722 - 23900787 on Build GRCm38
UniGene ID
Mm.436677
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039238.2 NP_001034327.2 4 491
GenBank mRNA BC059096.1 - 3 510
BC171996.1 - 492 9054282
DQ237937.1 - 491

View Full Product Details