Gene Transcripts

Gene Symbol
Grm4
Entrez Gene ID
24417
Gene Name
glutamate receptor, metabotropic 4
Gene Aliases
Chromosome Location
Chr. 20: 6745682 - 6791521 on Build Rnor_6.0
UniGene ID
Rn.89046
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022666.1 NP_073157.1 2253
GenBank mRNA M90518.1 - 2253
M92077.1 - 1468
U47331.1 - 2253

View Full Product Details