Gene Transcripts
- Gene Symbol
- Smc1a
- Entrez Gene ID
- 63996
- Gene Name
- structural maintenance of chromosomes 1A
- Gene Aliases
- SB1.8, SMC-1A, Smc1l1
- Chromosome Location
- Chr. X: 21710976 - 21755708 on Build Rnor_6.0
- UniGene ID
- Rn.11763
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031683.1 |
NP_113871.1
|
18 |
2896 |
|
GenBank mRNA |
AJ005113.1 |
- |
18 |
2896 |
|
View Full Product Details