Gene Transcripts

Gene Symbol
Thoc3
Entrez Gene ID
290519
Gene Name
THO complex 3
Gene Aliases
Chromosome Location
Chr. 17: 10676943 - 10686134 on Build Rnor_6.0
UniGene ID
Rn.101956
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106059.1 NP_001099529.1 4 715
GenBank mRNA BC168852.1 - 4 698

View Full Product Details