Gene Transcripts

Gene Symbol
Hdac1
Entrez Gene ID
297893
Gene Name
histone deacetylase 1
Gene Aliases
Chromosome Location
Chr. 5: 147716664 - 147743723 on Build Rnor_6.0
UniGene ID
Rn.1863
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025409.1 NP_001020580.1 6 545
GenBank mRNA BC059156.1 - 6 562
BC097943.1 - 6 545 7458532
BC107476.1 - 6 542 7443793

View Full Product Details