Gene Transcripts

Gene Symbol
Olr1252
Entrez Gene ID
300577
Gene Name
olfactory receptor 1252
Gene Aliases
Chromosome Location
Chr. 8: 42364887 - 42365819 on Build Rnor_6.0
UniGene ID
Rn.143518
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000454.1 NP_001000454.1 1 520

Gene Symbol
LOC100909736
Entrez Gene ID
100909736
Gene Name
olfactory receptor 8B3-like
Gene Aliases
Chromosome Location
Chr. 8: 42360580 - 42361272 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750485.1 XP_003750533.1 331

Gene Symbol
LOC100912507
Entrez Gene ID
100912507
Gene Name
olfactory receptor 8B3-like
Gene Aliases
Chromosome Location
Chr. 3: 158919862 - 158920794 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762440.1 XP_008760662.1 1 520

Gene Symbol
LOC103692018
Entrez Gene ID
103692018
Gene Name
olfactory receptor 8B3-like
Gene Aliases
Chromosome Location
Chr. 4: 1685913 - 1686845 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762660.1 XP_008760882.1 1 520

View Full Product Details