Gene Transcripts

Gene Symbol
Rnase10
Entrez Gene ID
305840
Gene Name
ribonuclease, RNase A family, 10 (non-active)
Gene Aliases
RGD1310415, Rah1
Chromosome Location
Chr. 15: 27930583 - 27933873 on Build Rnor_6.0
UniGene ID
Rn.215812
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012467.1 NP_001012485.1 1 202
XM_006251868.2 XP_006251930.1 2 1709

View Full Product Details