Gene Transcripts

Gene Symbol
Vgll2
Entrez Gene ID
309772
Gene Name
vestigial-like family member 2
Gene Aliases
Chromosome Location
Chr. 20: 33077106 - 33082962 on Build Rnor_6.0
UniGene ID
Rn.17643
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107630.1 NP_001101100.1 3 591
XM_006256448.2 XP_006256510.1 3 584
XM_008772885.1 XP_008771107.1 3 1204

View Full Product Details