Gene Transcripts

Gene Symbol
Naa15
Entrez Gene ID
310399
Gene Name
N(alpha)-acetyltransferase 15, NatA auxiliary subunit
Gene Aliases
Narg1
Chromosome Location
Chr. 2: 140471690 - 140534259 on Build Rnor_6.0
UniGene ID
Rn.12587
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107674.1 NP_001101144.1 20 2776

View Full Product Details